Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016403.3 | NP_057487.2 | 4, 5 | 467 | ||
XM_011542868.2 | XP_011541170.1 | 4, 5 | 602 | |||
XM_017017916.1 | XP_016873405.1 | 4, 5 | 655 | |||
GenBank mRNA | AF110775.1 | - | 4, 5 | 407 | ||
AF161497.1 | - | 4, 5 | 407 | |||
AJ250393.1 | - | 4, 5 | 424 | |||
AK225956.1 | - | 4, 5 | 426 | |||
AK314900.1 | - | 4, 5 | 411 | |||
BC006975.1 | - | 4, 5 | 430 | |||
BC032629.1 | - | 4, 5 | 425 | |||
BC040946.1 | - | 4, 5 | 419 | 6067745 | ||
BP231785.1 | - | 4, 5 | 467 |