Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 17 | 2837 | ||
XM_005260421.2 | XP_005260478.1 | 14 | 2447 | |||
XM_011528843.1 | XP_011527145.1 | 16 | 2438 | |||
XM_011528844.1 | XP_011527146.1 | 14 | 2509 | |||
XM_017027864.1 | XP_016883353.1 | 18 | 2919 | |||
XM_017027865.1 | XP_016883354.1 | 18 | 3120 | |||
XM_017027866.1 | XP_016883355.1 | 17 | 3040 | |||
XM_017027867.1 | XP_016883356.1 | 17 | 2875 | |||
XM_017027868.1 | XP_016883357.1 | 15 | 2661 | |||
XM_017027869.1 | XP_016883358.1 | 17 | 2834 | |||
XM_017027870.1 | XP_016883359.1 | 17 | 2840 | |||
XM_017027871.1 | XP_016883360.1 | 18 | 2920 | |||
GenBank mRNA | AK097498.1 | - | 6 | 1040 | ||
AK314503.1 | - | 17 | 2801 | |||
AY027523.1 | - | 16 | 2762 | |||
BC150178.1 | - | 17 | 2803 | 40148074 |