Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016441.2 | NP_057525.1 | 17 | 5011 | ||
XM_005264357.4 | XP_005264414.1 | 16 | 4833 | |||
XM_011532898.2 | XP_011531200.1 | 18 | 5132 | |||
XM_011532899.2 | XP_011531201.1 | 17 | 4938 | |||
XM_011532901.2 | XP_011531203.1 | 17 | 4911 | |||
XM_017004258.1 | XP_016859747.1 | 17 | 4889 | |||
XM_017004259.1 | XP_016859748.1 | 16 | 4955 | |||
XM_017004260.1 | XP_016859749.1 | 16 | 4764 | |||
XM_017004261.1 | XP_016859750.1 | 16 | 4424 | |||
XM_017004262.1 | XP_016859751.1 | 15 | 4307 | |||
GenBank mRNA | AF167706.1 | - | 17 | 4984 | ||
BC026227.1 | - | 1 | 1638 | |||
BX648755.1 | - | 11 | 3656 | |||
CR627451.1 | - | 2 | 3494 |