Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288564.1 | NP_001275493.1 | 6 | 1548 | ||
NM_001288565.1 | NP_001275494.1 | 5 | 2051 | |||
NM_001288566.1 | NP_001275495.1 | 6 | 1649 | |||
NM_001288567.1 | NP_001275496.1 | 6 | 1863 | |||
NM_001288568.1 | NP_001275497.1 | 6 | 1701 | |||
NM_001288569.1 | NP_001275498.1 | 6 | 1562 | |||
NM_001288570.1 | NP_001275499.1 | 6 | 1575 | |||
NM_001288571.1 | NP_001275500.1 | 6 | 1641 | |||
NM_016456.4 | NP_057540.1 | 6 | 1566 | |||
NR_109920.1 | 7 | 1793 | ||||
NR_109921.1 | 5 | 2020 | ||||
XM_011509385.1 | XP_011507687.1 | 6 | 1625 | |||
XM_011509386.2 | XP_011507688.1 | 6 | 1600 | |||
XM_011509387.2 | XP_011507689.1 | 6 | 1488 | |||
GenBank mRNA | AF151020.1 | - | 7 | 1539 | ||
AK054883.1 | - | 6 | 1521 | |||
AK075335.1 | - | 6 | 1476 | |||
AK091726.1 | - | 5 | 1936 | |||
AK094588.1 | - | 2 | 2869 | |||
AL136600.1 | - | 6 | 1505 | |||
AY359012.1 | - | 6 | 1540 | |||
BC001106.2 | - | 6 | 1491 | 3506442 |