Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128150.1 | NP_001121622.1 | 4 | 630 | ||
NM_016467.4 | NP_057551.1 | 5 | 740 | |||
XM_005246970.2 | XP_005247027.1 | 4 | 659 | |||
XM_017005347.1 | XP_016860836.1 | 5 | 779 | |||
GenBank mRNA | AB064959.1 | - | 3 | 1199 | ||
AB064960.1 | - | 4 | 638 | |||
AF151036.1 | - | 4 | 578 | |||
AF395704.1 | - | 5 | 695 | |||
AK074756.1 | - | 4 | 629 | |||
AK075160.1 | - | 3 | 631 | |||
AK126336.1 | - | 1 | 3665 | |||
BC005200.1 | - | 4 | 597 | 3929844 | ||
CR936635.1 | - | 3 | 1307 |