Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204090.1 | NP_001191019.1 | 3 | 553 | ||
NM_016468.6 | NP_057552.1 | 4 | 625 | |||
GenBank mRNA | AF151037.1 | - | 4 | 572 | ||
AF226729.1 | - | 4 | 599 | |||
AK290743.1 | - | 4 | 574 | |||
BC001702.2 | - | 4 | 526 | 2966935 | ||
CB156527.1 | - | 3 | 497 | |||
FJ460510.1 | - | 4 | 625 | |||
FJ460511.1 | - | 4 | 625 |
Gene Symbol: SYNJ2BP-COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202547.2 | NP_001189476.1 | 6 | 863 | ||
NM_001202548.2 | NP_001189477.1 | 6 | 836 | |||
NM_001202549.2 | NP_001189478.1 | 5 | 764 |