Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JKAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098625.2 | NP_001092095.1 | 7 | 1276 | ||
NM_001284201.1 | NP_001271130.1 | 7 | 1817 | |||
NM_001284202.1 | NP_001271131.1 | 8 | 1403 | |||
NM_001284203.1 | NP_001271132.1 | 7 | 1326 | |||
NM_001284204.1 | NP_001271133.1 | 6 | 1217 | |||
NM_016475.4 | NP_057559.2 | 7 | 1294 | |||
XM_006720171.3 | XP_006720234.1 | 6 | 1193 | |||
XM_017021366.1 | XP_016876855.1 | 6 | 1126 | |||
GenBank mRNA | AF151047.1 | - | 7 | 1216 | ||
AF161445.1 | - | 7 | 1215 | |||
AF212245.1 | - | 7 | 1216 | |||
AF529304.1 | - | 5 | 1531 | |||
AK021683.1 | - | 7 | 1175 | |||
AK056145.1 | - | 7 | 1693 | |||
AK128628.1 | - | 1 | 3122 | |||
AL136696.1 | - | 7 | 1186 | |||
BC005198.1 | - | 7 | 1202 | 3838401 | ||
BC010359.1 | - | 7 | 1233 | 4109351 | ||
BX161501.1 | - | 8 | 1286 |
Set Membership: |
G-protein coupled receptor |