Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000767.4 | NP_000758.1 | 9 | 2785 | ||
XM_011526548.2 | XP_011524850.1 | 6 | 2398 | |||
XM_011526549.2 | XP_011524851.1 | 7 | 2340 | |||
XM_011526550.2 | XP_011524852.1 | 5 | 2193 | |||
GenBank mRNA | M29874.1 | - | 9 | 2783 |