Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171201.1 | NP_001164672.1 | 2 | 283 | ||
NM_001171203.2 | NP_001164674.1 | 4 | 479 | |||
NM_001171204.2 | NP_001164675.1 | 4 | 447 | |||
NM_016525.4 | NP_057609.2 | 3 | 324 | |||
NR_033243.2 | 4 | 440 | ||||
XM_006716779.3 | XP_006716842.1 | 3 | 350 | |||
XM_011517899.1 | XP_011516201.1 | 4 | 381 | |||
XM_017014801.1 | XP_016870290.1 | 4 | 716 | |||
GenBank mRNA | AF222043.2 | - | 3 | 253 | ||
AK074426.1 | - | 3 | 317 | |||
AK074724.1 | - | 1 | 62 | |||
AK074745.1 | - | 3 | 284 | |||
AK074812.1 | - | 3 | 324 | |||
AK074969.1 | - | 4 | 426 | |||
AK074995.1 | - | 3 | 288 | |||
AK223235.1 | - | 3 | 279 | |||
AK303711.1 | - | 2 | 283 | |||
AL136733.1 | - | 3 | 117 | |||
BC020950.1 | - | 3 | 286 | 3851813 | ||
BC098141.1 | - | 3 | 220 | 40011247 | ||
BC098316.1 | - | 3 | 220 | 40011248 | ||
BC099726.1 | - | 3 | 220 | 40011249 | ||
BC100668.1 | - | 3 | 220 | 40011243 | ||
BP237024.1 | - | 4 | 408 | |||
BX473128.1 | - | 4 | 432 | |||
CR533551.1 | - | 2 | 81 | |||
DA741347.1 | - | 4 | 434 |