Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000772.2 | NP_000763.1 | 9 | 1894 | ||
NM_001128925.1 | NP_001122397.1 | 8 | 1717 | |||
GenBank mRNA | BC069666.1 | - | 9 | 1762 | 7262569 | |
BC096257.1 | - | 9 | 1613 | 40006072 | ||
BC096258.1 | - | 9 | 1613 | 40006073 | ||
BC096260.1 | - | 8 | 1436 | 40006076 | ||
BX538127.1 | - | 6 | 1028 | |||
M61853.1 | - | 9 | 1610 | |||
M61856.1 | - | 9 | 1767 |