Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193285.2 | NP_001180214.1 | 7 | 1521 | ||
NM_001321058.1 | NP_001307987.1 | 7 | 1474 | |||
NM_001321059.1 | NP_001307988.1 | 7 | 1419 | |||
NM_001321060.1 | NP_001307989.1 | 8 | 1562 | |||
NM_001321061.1 | NP_001307990.1 | 6 | 1393 | |||
NM_001321062.1 | NP_001307991.1 | 6 | 1291 | |||
NM_001321063.1 | NP_001307992.1 | 7 | 1379 | |||
NM_001321064.1 | NP_001307993.1 | 8 | 1527 | |||
NM_016539.3 | NP_057623.2 | 8 | 1602 | |||
GenBank mRNA | AF233396.1 | - | 8 | 1597 | ||
AK074810.1 | - | 8 | 1565 | |||
AK293348.1 | - | 7 | 1435 | |||
AL365375.1 | - | 993 | ||||
BC004218.2 | - | 6 | 1333 | 3352203 | ||
BC005026.1 | - | 8 | 1540 | 2820472 | ||
BC028220.1 | - | 7 | 1467 | 5217599 |