Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016580.3 | NP_057664.1 | 1 | 1333 | ||
GenBank mRNA | AB026893.1 | - | 1 | 179 | ||
AF231025.1 | - | 1 | 428 | |||
AF240635.1 | - | 1 | 1333 | |||
AK024140.1 | - | 1 | 471 | |||
AK027282.1 | - | 1 | 424 | |||
AK226049.1 | - | 1 | 430 | |||
AK315029.1 | - | 1 | 430 | |||
AY358428.1 | - | 1 | 450 | |||
BC042634.1 | - | 1 | 383 | 5177606 | ||
BC052973.1 | - | 1 | 440 | 5174442 |