Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278710.1 | NP_001265639.1 | 3 | 269 | ||
NM_001278711.1 | NP_001265640.1 | 3 | 508 | |||
NM_016588.2 | NP_057672.1 | 2 | 767 | |||
XM_006715106.3 | XP_006715169.1 | 4 | 1158 | |||
XM_006715107.3 | XP_006715170.1 | 2 | 948 | |||
GenBank mRNA | AF114833.1 | - | 2 | 298 | ||
AF136631.1 | - | 2 | 284 | |||
AJ420483.1 | - | 2 | 241 | |||
AK093824.1 | - | 2 | 962 | |||
AK314095.1 | - | 2 | 320 | |||
AL541038.3 | - | 2 | 353 | |||
BC002683.2 | - | 2 | 245 | 3605775 | ||
BC042019.1 | - | 2 | 768 | 5312094 | ||
BF970432.1 | - | 2 | 160 | |||
BX340783.2 | - | 3 | 506 | |||
DB013243.1 | - | 3 | 269 |