Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286703.1 | NP_001273632.1 | 6 | 485 | ||
NM_001286704.1 | NP_001273633.1 | 6 | 663 | |||
NM_001286705.1 | NP_001273634.1 | 7 | 527 | |||
NM_001286706.1 | NP_001273635.1 | 6 | 1161 | |||
NM_016617.3 | NP_057701.1 | 6 | 491 | |||
NR_104584.1 | 7 | 552 | ||||
NR_104585.1 | 5 | 451 | ||||
GenBank mRNA | AF208844.1 | - | 6 | 381 | ||
AK095490.1 | - | 6 | 411 | |||
AL515797.3 | - | 5 | 343 | |||
BC005193.1 | - | 6 | 408 | 3829206 | ||
BG533062.1 | - | 6 | 586 | |||
BG540094.1 | - | 7 | 414 | |||
BG717690.1 | - | 5 | 375 | |||
BI458294.1 | - | 6 | 461 | |||
BI761948.1 | - | 6 | 368 | |||
DA448931.1 | - | 5 | 364 | |||
DA483023.1 | - | 6 | 411 |