Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000782.4 | NP_000773.2 | 8 | 1521 | ||
NM_001128915.1 | NP_001122387.1 | 8 | 1521 | |||
XM_005260304.4 | XP_005260361.1 | 8 | 1529 | |||
XM_017027691.1 | XP_016883180.1 | 8 | 1529 | |||
XM_017027692.1 | XP_016883181.1 | 8 | 1529 | |||
XM_017027693.1 | XP_016883182.1 | 8 | 1529 | |||
GenBank mRNA | AY858838.1 | - | 6 | 957 | ||
BC109083.1 | - | 8 | 1208 | 40034385 | ||
BC109084.1 | - | 8 | 1208 | 40034387 | ||
L13286.1 | - | 8 | 1513 |