Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007367.3 | NP_031393.2 | 3 | 745 | ||
NM_016732.2 | NP_057951.1 | 3 | 745 | |||
XM_005260334.4 | XP_005260391.1 | 2 | 660 | |||
XM_005260336.4 | XP_005260393.1 | 2 | 660 | |||
XM_011528694.2 | XP_011526996.1 | 3 | 303 | |||
XM_011528695.2 | XP_011526997.1 | 4 | 855 | |||
XM_017027731.1 | XP_016883220.1 | 2 | 660 | |||
GenBank mRNA | AF148457.1 | - | 3 | 471 | ||
AK222915.1 | - | 3 | 431 | |||
AK226085.1 | - | 3 | 644 | |||
AK291054.1 | - | 3 | 316 | |||
AK291079.1 | - | 3 | 322 | |||
AK299766.1 | - | 3 | 486 | |||
BC103753.1 | - | 3 | 492 | 6182358 | ||
BC105018.1 | - | 3 | 340 | 8144021 | ||
L38696.1 | - | 3 | 349 |