Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032731.1 | NP_001027903.1 | 2 | 412 | ||
NM_002535.2 | NP_002526.2 | 2 | 412 | |||
NM_016817.2 | NP_058197.2 | 2 | 412 | |||
GenBank mRNA | AK292796.1 | - | 2 | 301 | ||
AK292906.1 | - | 2 | 329 | |||
BC010625.1 | - | 2 | 398 | |||
BC023637.2 | - | 2 | 333 | |||
BC049215.1 | - | 2 | 313 | 5517804 | ||
BC053870.1 | - | 2 | 335 | |||
BC069052.1 | - | 2 | 324 | |||
BE885222.1 | - | 469 | ||||
M87284.1 | - | 2 | 291 | |||
M87434.1 | - | 2 | 291 |