Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005783.2 | NP_001005783.2 | 5 | 794 | ||
NM_016527.3 | NP_057611.1 | 4 | 574 | |||
XM_011541558.1 | XP_011539860.1 | 3 | 372 | |||
XM_011541559.1 | XP_011539861.1 | 3 | 447 | |||
XM_011541560.1 | XP_011539862.1 | 3 | 447 | |||
XM_011541561.1 | XP_011539863.1 | 3 | 447 | |||
XM_017001421.1 | XP_016856910.1 | 5 | 1130 | |||
GenBank mRNA | AF203975.1 | - | 5 | 701 | ||
AF231917.1 | - | 4 | 512 | |||
AK298289.1 | - | 5 | 794 | |||
AY513277.1 | - | 4 | 479 | |||
BC020863.1 | - | 5 | 761 | 4557798 | ||
CB121424.1 | - | 4 | 496 |