Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000849.4 | NP_000840.2 | 9 | 1333 | ||
NR_024537.1 | 8 | 1257 | ||||
GenBank mRNA | AB209816.1 | - | 8 | 1042 | ||
AK304300.1 | - | 7 | 985 | |||
BC000088.2 | - | 8 | 1038 | 3509784 | ||
J05459.1 | - | 8 | 1041 |