Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016578.3 | NP_057662.3 | 6 | 1076 | ||
XM_005274051.2 | XP_005274108.1 | 6 | 1111 | |||
XM_017017923.1 | XP_016873412.1 | 6 | 850 | |||
GenBank mRNA | AF059317.1 | - | 4 | 627 | ||
AF227948.2 | - | 1 | 1261 | |||
AF380176.1 | - | 6 | 947 | |||
BC046124.1 | - | 2 | 359 | |||
BC046347.1 | - | 267 |