Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECSIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142464.2 | NP_001135936.1 | 6 | 959 | ||
NM_001142465.2 | NP_001135937.1 | 5 | 466 | |||
NM_001243204.1 | NP_001230133.1 | 7 | 997 | |||
NM_016581.4 | NP_057665.2 | 7 | 1108 | |||
GenBank mRNA | AF243044.1 | - | 6 | 1007 | ||
AK314905.1 | - | 7 | 997 | |||
BC000193.1 | - | 7 | 1056 | 3352568 | ||
BC005119.1 | - | 7 | 1051 | 2819610 | ||
BC008279.1 | - | 6 | 924 | 3639801 | ||
BE791936.1 | - | 5 | 467 | |||
BU186092.1 | - | 4 | 239 | |||
CR457204.1 | - | 6 | 974 |