Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271803.1 | NP_001258732.1 | 6, 7 | 774 | ||
NM_016606.3 | NP_057690.2 | 6, 7 | 768 | |||
NR_073448.1 | 6, 7 | 843 | ||||
NR_073449.1 | 6, 7 | 849 | ||||
GenBank mRNA | AF070551.1 | - | 2, 3 | 212 | ||
AF251041.1 | - | 5, 6 | 548 | |||
AK056193.1 | - | 6, 7 | 730 | |||
AK223611.1 | - | 6, 7 | 715 | |||
AK290547.1 | - | 6, 7 | 664 | |||
AK291101.1 | - | 6, 7 | 727 | |||
AK293480.1 | - | 6, 7 | 742 | |||
AY562240.1 | - | 6, 7 | 560 | |||
BC006218.2 | - | 6, 7 | 682 | 3940709 |