Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016614.2 | NP_057698.2 | 4, 5 | 550 | ||
GenBank mRNA | AF201687.1 | - | 4, 5 | 550 | ||
AF223469.1 | - | 4, 5 | 541 | |||
AJ269473.1 | - | 4, 5 | 546 | |||
AJ420495.1 | - | 3, 4 | 751 | |||
AK002168.1 | - | 4, 5 | 525 | |||
AK296623.1 | - | 3, 4 | 226 | |||
AK298699.1 | - | 4, 5 | 755 | |||
AK308836.1 | - | 4, 5 | 553 | |||
BC017553.2 | - | 4, 5 | 562 | 3920790 | ||
BC110375.1 | - | 4, 5 | 531 | 5737150 |