Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016622.3 | NP_057706.2 | 3, 4 | 437 | ||
NM_145644.2 | NP_663619.1 | 3, 4 | 437 | |||
XM_005264361.2 | XP_005264418.1 | 3, 4 | 515 | |||
GenBank mRNA | AF208849.1 | - | 3, 4 | 397 | ||
AK074352.1 | - | 3, 4 | 409 | |||
AK314555.1 | - | 3, 4 | 407 | |||
AK314577.1 | - | 3, 4 | 407 | |||
AL832982.1 | - | 3, 4 | 385 | |||
BC020651.1 | - | 3, 4 | 413 | 4709909 | ||
BQ421154.1 | - | 2, 3 | 274 | |||
DA413384.1 | - | 3, 4 | 515 | |||
DA443416.1 | - | 3, 4 | 413 |