Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTA4H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000895.2 | NP_000886.1 | 12 | 1308 | ||
NM_001256643.1 | NP_001243572.1 | 12 | 1239 | |||
NM_001256644.1 | NP_001243573.1 | 12 | 1239 | |||
NR_132659.1 | 12 | 1247 | ||||
XM_005268871.1 | XP_005268928.1 | 12 | 1298 | |||
XM_011538348.1 | XP_011536650.1 | 12 | 1298 | |||
XM_011538349.2 | XP_011536651.1 | 12 | 1298 | |||
GenBank mRNA | AB209740.1 | - | 13 | 1244 | ||
AK293630.1 | - | 12 | 1227 | |||
AK298017.1 | - | 12 | 1239 | |||
AK301308.1 | - | 12 | 1117 | |||
AK308184.1 | - | 12 | 1237 | |||
AK310461.1 | - | 2 | 202 | |||
BC032528.1 | - | 12 | 1225 | 5502553 | ||
BX647158.1 | - | 12 | 1196 | |||
CR457068.1 | - | 12 | 1166 | |||
J02959.1 | - | 12 | 1196 | |||
J03459.1 | - | 12 | 1234 |