Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267039.1 | NP_001253968.1 | 12, 13 | 1897 | ||
NM_015454.2 | NP_056269.1 | 10, 11 | 1561 | |||
NM_016648.3 | NP_057732.2 | 10, 11 | 1689 | |||
NR_049768.1 | 10, 11 | 1586 | ||||
GenBank mRNA | AF068284.1 | - | 9, 10 | 838 | ||
AK000274.1 | - | 10, 11 | 1499 | |||
AK225900.1 | - | 7, 8 | 1058 | |||
AL049996.1 | - | 4, 5 | 536 | |||
BC066945.1 | - | 12, 13 | 1735 | 4837780 | ||
BC107709.1 | - | 4, 5 | 530 | |||
BX647100.1 | - | 10, 11 | 1423 | |||
EU667388.1 | - | 10, 11 | 1432 |