Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PER3 SIRNA Assays ›
Gene Symbol:
PER3
Gene Name:
period circadian clock 3
Gene Aliases:
FASPS3, GIG13
Chromosome Location:
Chr.1: 7784285 - 7845181 on Build GRCh38
UniGene ID:
Hs.162200
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24762
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PER3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289861.1
NP_001276790.1
5
700
NM_001289862.1
NP_001276791.1
5
700
NM_001289863.1
NP_001276792.1
5
700
NM_001289864.1
NP_001276793.1
5
700
NM_016831.2
NP_058515.1
5
700
XM_005263524.3
XP_005263581.1
4
575
XM_011542385.2
XP_011540687.1
4
575
XM_011542390.2
XP_011540692.1
4
575
XM_017002723.1
XP_016858212.1
4
575
XM_017002724.1
XP_016858213.1
4
575
XM_017002725.1
XP_016858214.1
4
575
XM_017002726.1
XP_016858215.1
4
575
XM_017002727.1
XP_016858216.1
6
894
XM_017002728.1
XP_016858217.1
4
636
XM_017002729.1
XP_016858218.1
2
79
XM_017002730.1
XP_016858219.1
4
636
XM_017002731.1
XP_016858220.1
4
636
XM_017002732.1
XP_016858221.1
3
685
XM_017002733.1
XP_016858222.1
6
801
XM_017002734.1
XP_016858223.1
7
943
XM_017002735.1
XP_016858224.1
6
801
XM_017002736.1
XP_016858225.1
7
943
XM_017002737.1
XP_016858226.1
2
186
XM_017002738.1
XP_016858227.1
2
192
GenBank mRNA
AB047686.1
-
4
660
AK307760.1
-
4
575
AK308303.1
-
5
749
AL157954.1
-
4
611
AY493418.1
-
4
507
BC026102.1
-
5
722
BC144137.1
-
4
604
BC146781.1
-
4
660
9007235
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011067, NM_001289877, NM_001289878, AK166514, BC145178, and others.
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
circadian regulation of gene expression
regulation of circadian sleep/wake cycle, sleep
protein stabilization
protein binding
kinase binding
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0