Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016530.2 | NP_057614.1 | 8 | 953 | ||
XM_017022312.1 | XP_016877801.1 | 9 | 1008 | |||
GenBank mRNA | AB038995.1 | - | 8 | 948 | ||
AK001111.1 | - | 8 | 876 | |||
AK095444.1 | - | 4 | 290 | |||
AK297598.1 | - | 7 | 825 | |||
AL833365.1 | - | 8 | 866 | |||
BC020654.1 | - | 8 | 865 | 4701429 |