Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000962.3 | NP_000953.2 | 11 | 4211 | ||
NM_001271164.1 | NP_001258093.1 | 10 | 4067 | |||
NM_001271165.1 | NP_001258094.1 | 10 | 4160 | |||
NM_001271166.1 | NP_001258095.1 | 11 | 4326 | |||
NM_001271367.1 | NP_001258296.1 | 11 | 4071 | |||
NM_001271368.1 | NP_001258297.1 | 12 | 4548 | |||
NM_080591.2 | NP_542158.1 | 11 | 4100 | |||
XM_005252105.3 | XP_005252162.1 | 12 | 4386 | |||
XM_011518875.2 | XP_011517177.1 | 12 | 4647 | |||
XM_011518876.2 | XP_011517178.1 | 12 | 8257 | |||
XM_017014929.1 | XP_016870418.1 | 14 | 4621 | |||
GenBank mRNA | AF129755.1 | - | 1 | 66 | ||
AJ420464.1 | - | 1 | 568 |