Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288998.1 | NP_001275927.1 | 20 | 3095 | ||
NM_001288999.1 | NP_001275928.1 | 20 | 3104 | |||
NM_016573.3 | NP_057657.2 | 21 | 3182 | |||
XM_005259927.2 | XP_005259984.1 | 21 | 3158 | |||
XM_011528048.2 | XP_011526350.1 | 20 | 3049 | |||
XM_017026861.1 | XP_016882350.1 | 21 | 3211 | |||
GenBank mRNA | AF086104.1 | - | 1 | 234 | ||
AF132541.1 | - | 21 | 3123 |