Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016577.3 | NP_057661.3 | 8 | 1120 | ||
XM_011512893.2 | XP_011511195.1 | 8 | 795 | |||
GenBank mRNA | AB073348.1 | - | 2 | 239 | ||
AF166492.1 | - | 8 | 1076 | |||
AK295451.1 | - | 9 | 857 | |||
BC002510.2 | - | 8 | 1089 | 3050592 | ||
BC078662.1 | - | 8 | 868 | 6450986 |