Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016585.4 | NP_057669.1 | 8 | 1298 | ||
NM_199202.2 | NP_954672.1 | 7 | 1226 | |||
XM_011528049.2 | XP_011526351.1 | 9 | 2073 | |||
XM_011528050.2 | XP_011526352.1 | 8 | 2000 | |||
XM_011528051.2 | XP_011526353.1 | 6 | 1870 | |||
GenBank mRNA | AF268610.1 | - | 8 | 1264 |