Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144891.1 | NP_001138363.1 | 7 | 933 | ||
NM_016586.2 | NP_057670.2 | 7 | 933 | |||
XM_005267754.3 | XP_005267811.1 | 6 | 862 | |||
GenBank mRNA | AB038523.1 | - | 7 | 902 | ||
AF208857.1 | - | 7 | 882 | |||
AK293473.1 | - | 7 | 814 | |||
AK315290.1 | - | 7 | 934 | |||
BC005197.1 | - | 7 | 892 | 3681897 | ||
BC016821.1 | - | 7 | 888 | 4097046 |