Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMMDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016589.3 | NP_057673.2 | 6 | 875 | ||
XM_017006556.1 | XP_016862045.1 | 2 | 469 | |||
GenBank mRNA | AF139077.1 | - | 6 | 848 | ||
AF210057.1 | - | 6 | 827 | |||
AK026250.1 | - | 5 | 765 | |||
AK225954.1 | - | 5 | 776 | |||
AL136622.1 | - | 6 | 850 | |||
AL390077.1 | - | 6 | 827 | |||
AL390090.1 | - | 6 | 802 | |||
AL390094.1 | - | 6 | 509 | |||
AY358633.1 | - | 6 | 828 | |||
BC012341.1 | - | 6 | 769 | 4576562 | ||
CR457158.1 | - | 6 | 662 | |||
CR533524.1 | - | 6 | 662 |