Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190997.2 | NP_001177926.1 | 11 | 1282 | ||
NM_016615.4 | NP_057699.2 | 13 | 1558 | |||
XM_006719008.3 | XP_006719071.1 | 8 | 1345 | |||
XM_011521012.2 | XP_011519314.1 | 13 | 1626 | |||
XM_011521013.1 | XP_011519315.1 | 14 | 1694 | |||
XM_011521014.1 | XP_011519316.1 | 12 | 1450 | |||
XM_017019842.1 | XP_016875331.1 | 9 | 1443 | |||
XM_017019843.1 | XP_016875332.1 | 9 | 1041 | |||
XM_017019845.1 | XP_016875334.1 | 9 | 943 | |||
GenBank mRNA | AK296127.1 | - | 11 | 1245 | ||
AK296209.1 | - | 14 | 1489 | |||
AK313511.1 | - | 13 | 1514 | |||
BC022392.1 | - | 13 | 1521 | 4612245 | ||
U76343.2 | - | 13 | 1403 |