Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271834.1 | NP_001258763.1 | 2 | 281 | ||
NM_001271838.1 | NP_001258767.1 | 2 | 307 | |||
NM_016625.3 | NP_057709.2 | 2 | 295 | |||
GenBank mRNA | AF208853.1 | - | 2 | 253 | ||
AK290334.1 | - | 2 | 275 | |||
AK300388.1 | - | 2 | 191 | |||
BC006982.1 | - | 2 | 244 | 3997840 | ||
BC010357.1 | - | 2 | 230 | 4249317 | ||
DA273191.1 | - | 2 | 297 |