Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007367.3 | NP_031393.2 | 9 | 1735 | ||
NM_016732.2 | NP_057951.1 | 10 | 1783 | |||
XM_005260334.4 | XP_005260391.1 | 9 | 1698 | |||
XM_005260336.4 | XP_005260393.1 | 8 | 1650 | |||
XM_011528694.2 | XP_011526996.1 | 10 | 1341 | |||
XM_011528695.2 | XP_011526997.1 | 11 | 1893 | |||
GenBank mRNA | AF148457.1 | - | 10 | 1509 | ||
AK222915.1 | - | 10 | 1472 | |||
AK226085.1 | - | 9 | 1637 | |||
AK291054.1 | - | 9 | 1309 | |||
AK291079.1 | - | 10 | 1363 | |||
BC103753.1 | - | 10 | 1533 | 6182358 | ||
L38696.1 | - | 9 | 1342 |