Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289861.1 | NP_001276790.1 | 23 | 5324 | ||
NM_001289862.1 | NP_001276791.1 | 22 | 5381 | |||
NM_001289863.1 | NP_001276792.1 | 23 | 5303 | |||
NM_001289864.1 | NP_001276793.1 | 22 | 5439 | |||
NM_016831.2 | NP_058515.1 | 22 | 5354 | |||
XM_005263524.3 | XP_005263581.1 | 21 | 5302 | |||
XM_011542385.2 | XP_011540687.1 | 21 | 5253 | |||
XM_011542390.2 | XP_011540692.1 | 21 | 5232 | |||
XM_017002724.1 | XP_016858213.1 | 21 | 5250 | |||
XM_017002730.1 | XP_016858219.1 | 21 | 5314 | |||
XM_017002731.1 | XP_016858220.1 | 21 | 5293 | |||
XM_017002735.1 | XP_016858224.1 | 23 | 5479 | |||
XM_017002738.1 | XP_016858227.1 | 18 | 4743 | |||
GenBank mRNA | AB047686.1 | - | 21 | 5341 | ||
AL157954.1 | - | 21 | 5265 | |||
BC111010.1 | - | 6 | 2568 | |||
BC146781.1 | - | 21 | 5341 | 9007235 |