Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109878.1 | NP_001103348.1 | 9 | 2307 | ||
NM_001109879.1 | NP_001103349.1 | 9 | 2303 | |||
NM_001303475.1 | NP_001290404.1 | 7 | 2061 | |||
NM_016954.2 | NP_058650.1 | 8 | 2200 | |||
XM_005262136.2 | XP_005262193.1 | 9 | 2335 | |||
XM_011530972.1 | XP_011529274.1 | 8 | 2940 | |||
GenBank mRNA | AY035371.1 | - | 8 | 2179 | ||
BC014194.2 | - | 8 | 2200 | 3952809 |