Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A4GALT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318038.1 | NP_001304967.1 | 3 | 473 | ||
NM_017436.5 | NP_059132.1 | 3 | 450 | |||
XM_005261644.2 | XP_005261701.1 | 3 | 755 | |||
XM_005261646.4 | XP_005261703.1 | 2 | 311 | |||
XM_005261647.2 | XP_005261704.1 | 3 | 285 | |||
XM_005261648.3 | XP_005261705.1 | 3 | 368 | |||
XM_006724265.3 | XP_006724328.1 | 3 | 696 | |||
XM_006724266.3 | XP_006724329.1 | 3 | 430 | |||
XM_011530233.2 | XP_011528535.1 | 2 | 637 | |||
XM_017028831.1 | XP_016884320.1 | 2 | 567 | |||
GenBank mRNA | AB037883.1 | - | 2 | 282 | ||
AJ245581.1 | - | 1 | 149 | |||
AK074388.1 | - | 1 | 49 | |||
AK312927.1 | - | 3 | 454 | |||
BC017068.2 | - | 3 | 369 | 3913851 | ||
BC055286.1 | - | 2 | 639 | 6161307 | ||
GU902275.1 | - | 2 | 240 | |||
GU902276.1 | - | 2 | 240 | |||
GU902277.1 | - | 3 | 312 | |||
HY140390.1 | - | 3 | 451 | |||
KU178649.1 | - | 1 | 149 | |||
KU178650.1 | - | 1 | 149 |