Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100420.1 | NP_001093890.1 | 3 | 312 | ||
NM_001100421.1 | NP_001093891.1 | 3 | 312 | |||
NM_017447.3 | NP_059143.3 | 3 | 312 | |||
GenBank mRNA | AF239726.1 | - | 3 | 265 | ||
AK023825.1 | - | 3 | 277 | |||
AK225123.1 | - | 3 | 283 | |||
AK314471.1 | - | 3 | 285 | |||
BC015468.2 | - | 2 | 228 | 3916462 | ||
BF739854.1 | - | 3 | 311 | |||
CR457317.1 | - | 2 | 221 | |||
CR749496.1 | - | 3 | 292 | |||
DA215855.1 | - | 3 | 285 | |||
DB066982.1 | - | 1 | 27 |