Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291780.1 | NP_001278709.1 | 1 | 344 | ||
NM_017495.5 | NP_059965.2 | 1 | 344 | |||
NM_183425.2 | NP_906270.1 | 1 | 344 | |||
XM_005260446.3 | XP_005260503.1 | 1 | 282 | |||
XM_011528885.2 | XP_011527187.1 | 1 | 276 | |||
GenBank mRNA | AF432218.1 | - | 1 | 340 | ||
BC018711.1 | - | 1 | 96 | 4852523 | ||
BM549757.1 | - | 339 | ||||
DB504051.1 | - | 1 | 345 | |||
DN998561.1 | - | 1 | 329 | |||
X75314.1 | - | 1 | 131 |