Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198681.1 | NP_001185610.1 | 5 | 462 | ||
NM_017526.4 | NP_059996.1 | 4 | 598 | |||
GenBank mRNA | AK074841.1 | - | 4 | 359 | ||
AK130096.1 | - | 1 | 892 | |||
AK309370.1 | - | 5 | 462 | |||
BC011027.2 | - | 3 | 306 | |||
BC056250.1 | - | 4 | 343 | 4328021 | ||
BI715133.1 | - | 3 | 275 | |||
CB140261.1 | - | 4 | 466 | |||
CR541647.1 | - | 4 | 321 | |||
CR541737.1 | - | 4 | 321 | |||
Y12670.1 | - | 4 | 391 |