Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001432.2 | NP_001423.1 | 5 | 2605 | ||
GenBank mRNA | BC136404.1 | - | 5 | 2562 | 9020391 | |
BC136405.1 | - | 5 | 2562 | 9020392 | ||
D30783.1 | - | 5 | 2604 |