Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278603.1 | NP_001265532.1 | 2 | 255 | ||
NM_017426.3 | NP_059122.2 | 2 | 255 | |||
NR_103780.1 | 2 | 255 | ||||
NR_103781.1 | 2 | 255 | ||||
NR_103782.1 | 4 | 440 | ||||
XM_006714237.1 | XP_006714300.1 | 3 | 232 | |||
XM_011532033.2 | XP_011530335.1 | 2 | 198 | |||
XM_011532034.2 | XP_011530336.1 | 2 | 642 | |||
XM_017008294.1 | XP_016863783.1 | 2 | 642 | |||
XM_017008295.1 | XP_016863784.1 | 2 | 642 | |||
GenBank mRNA | AF157322.1 | - | 2 | 136 | ||
AK001517.1 | - | 2 | 137 | |||
AK223352.1 | - | 2 | 138 | |||
AK299442.1 | - | 2 | 137 | |||
AK300036.1 | - | 2 | 137 | |||
AK307571.1 | - | 4 | 330 | |||
AK315160.1 | - | 2 | 135 | |||
BC012559.1 | - | 2 | 152 | |||
BX538002.1 | - | 2 | 155 | |||
DA577078.1 | - | 2 | 255 |