Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001561.5 | NP_001552.2 | 9 | 1592 | ||
XM_011541386.2 | XP_011539688.1 | 8 | 1607 | |||
GenBank mRNA | BC006196.2 | - | 8 | 1452 | 2924109 |