Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017444.5 | NP_059140.1 | 2 | 411 | ||
NR_040712.1 | 2 | 106 | ||||
GenBank mRNA | AF226076.1 | - | 2 | 209 | ||
AK023537.1 | - | 2 | 404 | |||
AK311595.1 | - | 2 | 172 | |||
BC015891.1 | - | 2 | 248 | 3908717 | ||
BU160855.1 | - | 2 | 109 | |||
DA647606.1 | - | 2 | 411 | |||
DA887467.1 | - | 2 | 411 |