Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100420.1 | NP_001093890.1 | 3 | 411 | ||
NM_001100421.1 | NP_001093891.1 | 3 | 411 | |||
NM_017447.3 | NP_059143.3 | 3 | 411 | |||
GenBank mRNA | AF239726.1 | - | 3 | 364 | ||
AK023825.1 | - | 3 | 376 | |||
AK314471.1 | - | 3 | 384 | |||
BC015468.2 | - | 2 | 327 | 3916462 | ||
BF739854.1 | - | 3 | 410 | |||
CR457317.1 | - | 2 | 320 | |||
DA215855.1 | - | 3 | 384 | |||
DB066982.1 | - | 1 | 126 |