Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003218.3 | NP_003209.2 | 1, 2 | 351 | ||
NM_017489.2 | NP_059523.2 | 1, 2 | 351 | |||
XM_005251291.3 | XP_005251348.1 | 1, 2 | 349 | |||
XM_005251292.3 | XP_005251349.1 | 1, 2 | 349 | |||
GenBank mRNA | AF003001.1 | - | 1, 2 | 341 | ||
BC029378.1 | - | 1, 2 | 334 | 4724068 | ||
BC071975.1 | - | 1, 2 | 327 | |||
U40705.1 | - | 1, 2 | 341 | |||
U74382.1 | - | 1, 2 | 326 |