Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017515.4 | NP_059985.2 | 1, 2 | 523 | ||
GenBank mRNA | AK023080.1 | - | 1, 2 | 120 | ||
AK128062.1 | - | 4, 5 | 692 | |||
AL833969.1 | - | 1, 2 | 126 | |||
BC039195.2 | - | 1, 2 | 263 | 3847013 | ||
BC048302.1 | - | 1, 2 | 129 | 5499821 | ||
DA840333.1 | - | 1, 2 | 509 | |||
DC352180.1 | - | 1, 2 | 523 |